SY3H3D6802FE Synthonix Corporation

6-(Methoxymethyl)picolinaldehyde

  • Cas Number : 890904-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/49286.html

Safety Information

MSDS https://synthonix.com/49286.html
Availability details for
Quantity
  • Availability
  • Add to Cart