SY3H3D68030C Synthonix Corporation

Methyl 3-amino-5-hydroxybenzoate

  • MDL number No MDL found
  • Cas Number : 67973-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/49328.html

Safety Information

MSDS https://synthonix.com/49328.html
Availability details for
Quantity
  • Availability
  • Add to Cart