SY3H3D680310 Synthonix Corporation

Methyl 6-(hydroxymethyl)picolinate

  • Cas Number : 39977-44-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49333.html

Safety Information

MSDS https://synthonix.com/49333.html
Availability details for
Quantity
  • Availability
  • Add to Cart