SY3H3D680311 Synthonix Corporation

2-Methoxy-3-methylisonicotinic acid

  • Cas Number : 1211581-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49334.html

Safety Information

MSDS https://synthonix.com/49334.html
Availability details for
Quantity
  • Availability
  • Add to Cart