SY3H3D680316 Synthonix Corporation

Methyl 3-acetamidothiophene-2-carboxylate

  • Cas Number : 22288-79-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49346.html

Safety Information

MSDS https://synthonix.com/49346.html
Availability details for
Quantity
  • Availability
  • Add to Cart