SY3H3D680317 Synthonix Corporation

(2-Methoxy-5-nitrophenyl)methanol

  • Cas Number : 5804-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49352.html

Safety Information

MSDS https://synthonix.com/49352.html
Availability details for
Quantity
  • Availability
  • Add to Cart