SY3H3D680319 Synthonix Corporation

Methyl 5-hydroxy-6-methoxynicotinate

  • MDL number No MDL found
  • Cas Number : 166742-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49355.html

Safety Information

MSDS https://synthonix.com/49355.html
Availability details for
Quantity
  • Availability
  • Add to Cart