SY3H3D68031A Synthonix Corporation

Methyl 5-hydroxy-6-methoxypicolinate

  • MDL number No MDL found
  • Cas Number : 1256791-78-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/49356.html

Safety Information

MSDS https://synthonix.com/49356.html
Availability details for
Quantity
  • Availability
  • Add to Cart