SY3H3D68031E Synthonix Corporation

4-[(Methylamino)methyl]pyrocatechol Hydrobromide

  • MDL number No MDL found
  • Cas Number : 1025423-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49486.html

Safety Information

MSDS https://synthonix.com/49486.html
Availability details for
Quantity
  • Availability
  • Add to Cart