SY3H3D680320 Synthonix Corporation

3-Methylbenzimidamide hydrochloride

  • MDL number No MDL found
  • Cas Number : 20680-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49488.html

Safety Information

MSDS https://synthonix.com/49488.html
Availability details for
Quantity
  • Availability
  • Add to Cart