SY3H3D680326 Synthonix Corporation

Methyl 2-(aminomethyl)pyridine-4-carboxylate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1072438-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49501.html

Safety Information

MSDS https://synthonix.com/49501.html
Availability details for
Quantity
  • Availability
  • Add to Cart