SY3H3D680327 Synthonix Corporation

4-Methanesulfonylbenzamidine hydrochloride

  • MDL number No MDL found
  • Cas Number : 5434-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link https://synthonix.com/49504.html

Safety Information

MSDS https://synthonix.com/49504.html
Availability details for
Quantity
  • Availability
  • Add to Cart