SY3H3D68032A Synthonix Corporation

N-Methyl-4-(methylsulfonyl)aniline hydrochloride

  • Cas Number : 1263378-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49530.html

Safety Information

MSDS https://synthonix.com/49530.html
Availability details for
Quantity
  • Availability
  • Add to Cart