SY3H3D68032D Synthonix Corporation

2-(5-Methoxypyridin-2-yl)ethanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1624262-52-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/49539.html

Safety Information

MSDS https://synthonix.com/49539.html
Availability details for
Quantity
  • Availability
  • Add to Cart