SY3H3D68032E Synthonix Corporation

(S)-1-(2-Methoxypyridin-4-yl)ethanamine hydrochloride

  • MDL number No MDL found
  • Cas Number : 1914157-93-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/49540.html

Safety Information

MSDS https://synthonix.com/49540.html
Availability details for
Quantity
  • Availability
  • Add to Cart