SY3H3D680331 Synthonix Corporation

4-(4-Methyl-4H-1,2,4-triazol-3-yl)piperidine hydrochloride

  • MDL number No MDL found
  • Cas Number : 297171-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49558.html

Safety Information

MSDS https://synthonix.com/49558.html
Availability details for
Quantity
  • Availability
  • Add to Cart