SY3H3D680335 Synthonix Corporation

(S)-Methyl 2-amino-2-cyclopentylacetate hydrochloride

  • MDL number No MDL found
  • Cas Number : 14328-62-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49573.html

Safety Information

MSDS https://synthonix.com/49573.html
Availability details for
Quantity
  • Availability
  • Add to Cart