SY3H3D680336 Synthonix Corporation

Methyl 2-amino-2-(tetrahydro-2H-pyran-4-yl)acetate hydrochloride

  • MDL number No MDL found
  • Cas Number : 1260637-54-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49575.html

Safety Information

MSDS https://synthonix.com/49575.html
Availability details for
Quantity
  • Availability
  • Add to Cart