SY3H3D680337 Synthonix Corporation

1-(4-(Methylamino)piperidin-1-yl)ethanone hydrochloride

  • MDL number No MDL found
  • Cas Number : 71879-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49578.html

Safety Information

MSDS https://synthonix.com/49578.html
Availability details for
Quantity
  • Availability
  • Add to Cart