SY3H3D680341 Synthonix Corporation

1-(5-Methylpyridin-2-yl)ethanamine dihydrochloride

  • MDL number MFCD08752624
  • Cas Number : 1187931-95-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49667.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49667.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart