SY3H3D680345 Synthonix Corporation

2-(5-Methylpyridin-2-yl)acetonitrile

  • MDL number No MDL found
  • Cas Number : 38203-08-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/48792.html

Safety Information

MSDS https://synthonix.com/48792.html
Availability details for
Quantity
  • Availability
  • Add to Cart