SY3H3D68034A Synthonix Corporation

5-Methoxy-2-methyl-2H-indazole

  • Cas Number : 541539-88-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49721.html

Safety Information

MSDS https://synthonix.com/49721.html
Availability details for
Quantity
  • Availability
  • Add to Cart