SY3H3D68034E Synthonix Corporation

(1-Methyl-1H-benzo[d]imidazol-6-yl)methanol

  • MDL number No MDL found
  • Cas Number : 181867-18-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49727.html

Safety Information

MSDS https://synthonix.com/49727.html
Availability details for
Quantity
  • Availability
  • Add to Cart