SY3H3D680350 Synthonix Corporation

5-Methoxy-3-methyl-1H-pyrrolo[3,2-b]pyridine

  • Cas Number : 138469-76-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49729.html

Safety Information

MSDS https://synthonix.com/49729.html
Availability details for
Quantity
  • Availability
  • Add to Cart