SY3H3D680351 Synthonix Corporation

4-Methyl-5-nitroindoline

  • MDL number MFCD26743710
  • Cas Number : 165250-68-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49732.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49732.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart