SY3H3D680352 Synthonix Corporation

4-Methyl-6-nitroindoline

  • Cas Number : 1588441-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49733.html

Safety Information

MSDS https://synthonix.com/49733.html
Availability details for
Quantity
  • Availability
  • Add to Cart