SY3H3D680353 Synthonix Corporation

4-Methyl-7-nitroindoline

  • Cas Number : 179176-31-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49734.html

Safety Information

MSDS https://synthonix.com/49734.html
Availability details for
Quantity
  • Availability
  • Add to Cart