SY3H3D680357 Synthonix Corporation

N-Methyl-1-(4-(trifluoromethyl)phenyl)methanamine

  • MDL number No MDL found
  • Cas Number : 90390-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/49787.html

Safety Information

MSDS https://synthonix.com/49787.html
Availability details for
Quantity
  • Availability
  • Add to Cart