SY3H3D68035D Synthonix Corporation

Methyl 2-chloro-4,6-dimethylnicotinate

  • MDL number No MDL found
  • Cas Number : 339151-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49816.html

Safety Information

MSDS https://synthonix.com/49816.html
Availability details for
Quantity
  • Availability
  • Add to Cart