SY3H3D68035E Synthonix Corporation

Methyl 2-(3-amino-4-chlorophenyl)acetate

  • MDL number No MDL found
  • Cas Number : 59833-69-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/49820.html

Safety Information

MSDS https://synthonix.com/49820.html
Availability details for
Quantity
  • Availability
  • Add to Cart