SY3H3D680360 Synthonix Corporation

Methyl 2-amino-2-(3-chlorophenyl)acetate hydrochloride

  • MDL number MFCD16661102
  • Cas Number : 1351586-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form White to off-white solid
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49824.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49824.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart