SY3H3D680362 Synthonix Corporation

Methyl 5-fluoro-2-(methylamino)benzoate

  • MDL number No MDL found
  • Cas Number : 924668-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49833.html

Safety Information

MSDS https://synthonix.com/49833.html
Availability details for
Quantity
  • Availability
  • Add to Cart