SY3H3D680369 Synthonix Corporation

Methyl 4-amino-5-iodo-2-methoxybenzoate

  • MDL number No MDL found
  • Cas Number : 137832-56-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49849.html

Safety Information

MSDS https://synthonix.com/49849.html
Availability details for
Quantity
  • Availability
  • Add to Cart