SY3H3D68037A Synthonix Corporation

4-Methoxy-3-methylbenzoic acid

  • MDL number MFCD00270114
  • Cas Number : 6880-04-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: White - Slightly pale yellow
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/49918.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/49918.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart