SY3H3D68037D Synthonix Corporation

Methyl 3-hydroxy-2-methylbenzoate

  • MDL number No MDL found
  • Cas Number : 55289-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://synthonix.com/49923.html

Safety Information

MSDS https://synthonix.com/49923.html
Availability details for
Quantity
  • Availability
  • Add to Cart