SY3H3D680381 Synthonix Corporation

4-((Methylsulfonyl)methyl)benzaldehyde

  • MDL number No MDL found
  • Cas Number : 156867-56-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/49939.html

Safety Information

MSDS https://synthonix.com/49939.html
Availability details for
Quantity
  • Availability
  • Add to Cart