SY3H3D680384 Synthonix Corporation

Methyl 3-hydroxy-5-methoxybenzoate

  • MDL number No MDL found
  • Cas Number : 19520-74-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/49943.html

Safety Information

MSDS https://synthonix.com/49943.html
Availability details for
Quantity
  • Availability
  • Add to Cart