SY3H3D68038B Synthonix Corporation

(S)-Methyl 2-amino-3-(5-chloropyridin-2-yl)propanoate hydrochloride

  • Cas Number : 1810074-86-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/49988.html

Safety Information

MSDS https://synthonix.com/49988.html
Availability details for
Quantity
  • Availability
  • Add to Cart