SY3H3D680390 Synthonix Corporation

Methyl 6-ethylpicolinate

  • Cas Number : 103931-19-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50036.html

Safety Information

MSDS https://synthonix.com/50036.html
Availability details for
Quantity
  • Availability
  • Add to Cart