SY3H3D680391 Synthonix Corporation

4-Methoxy-N-methylbenzamide

  • Cas Number : 3400-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link https://synthonix.com/50044.html

Safety Information

MSDS https://synthonix.com/50044.html
Availability details for
Quantity
  • Availability
  • Add to Cart