SY3H3D680393 Synthonix Corporation

2-(4-Methoxyphenyl)acetamide

  • MDL number No MDL found
  • Cas Number : 6343-93-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://synthonix.com/50047.html

Safety Information

MSDS https://synthonix.com/50047.html
Availability details for
Quantity
  • Availability
  • Add to Cart