SY3H3D680394 Synthonix Corporation

N-(4-Methoxybenzyl)formamide

  • MDL number No MDL found
  • Cas Number : 17061-63-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50050.html

Safety Information

MSDS https://synthonix.com/50050.html
Availability details for
Quantity
  • Availability
  • Add to Cart