SY3H3D680397 Synthonix Corporation

Methyl 2-amino-5-methylbenzoate

  • Cas Number : 18595-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50059.html

Safety Information

MSDS https://synthonix.com/50059.html
Availability details for
Quantity
  • Availability
  • Add to Cart