SY3H3D68039A Synthonix Corporation

Methyl 5-amino-2-methylbenzoate

  • MDL number MFCD08752568
  • Cas Number : 18595-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. refrigerator
Physical Form Crystal - Powder / Colour: Yellow red - Deep yellow red - Red-brown
Shipping Temperature Room Temperature
Country of Origin USA
Product Link https://synthonix.com/50062.html

Safety Information

signalword Warning
pcodes P261
MSDS https://synthonix.com/50062.html
pictograms GHS07
hstatements H302, H315, H319, H335
Availability details for
Quantity
  • Availability
  • Add to Cart