SY3H3D68039B Synthonix Corporation

Methyl 2,6-dimethylisonicotinate

  • MDL number No MDL found
  • Cas Number : 142896-15-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://synthonix.com/50068.html

Safety Information

MSDS https://synthonix.com/50068.html
Availability details for
Quantity
  • Availability
  • Add to Cart