SY3H3D68039C Synthonix Corporation

Methyl 2-(4-methylpyridin-2-yl)acetate

  • MDL number No MDL found
  • Cas Number : 59310-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50073.html

Safety Information

MSDS https://synthonix.com/50073.html
Availability details for
Quantity
  • Availability
  • Add to Cart