SY3H3D68039D Synthonix Corporation

Methyl 2-(6-methylpyridin-3-yl)acetate

  • MDL number No MDL found
  • Cas Number : 90610-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link https://synthonix.com/50074.html

Safety Information

MSDS https://synthonix.com/50074.html
Availability details for
Quantity
  • Availability
  • Add to Cart