SY3H3D6803A4 Synthonix Corporation

Methyl 4-amino-3-(hydroxymethyl)benzoate

  • Cas Number : 76143-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50099.html

Safety Information

MSDS https://synthonix.com/50099.html
Availability details for
Quantity
  • Availability
  • Add to Cart