SY3H3D6803A6 Synthonix Corporation

1-(2-Methoxyethoxy)-4-nitrobenzene

  • MDL number No MDL found
  • Cas Number : 22483-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50108.html

Safety Information

MSDS https://synthonix.com/50108.html
Availability details for
Quantity
  • Availability
  • Add to Cart