SY3H3D6803AC Synthonix Corporation

2-Methyl-5,6,7,8-tetrahydroquinazolin-4-ol

  • MDL number No MDL found
  • Cas Number : 19178-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link https://synthonix.com/50175.html

Safety Information

MSDS https://synthonix.com/50175.html
Availability details for
Quantity
  • Availability
  • Add to Cart